EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Preparation of cocrystal nanofibres of cobalt octaethylporphyrin and tetracyanoquinodimethane with good photoresponse.
PMID 24738357
Abstract
Cocrystal nanofibres of cobalt octaethylporphyrin and tetracyanoquinodimethane were prepared by a facile solution method and fully characterized by SEM, AFM, XRD, Raman, EDX, and UV-vis-NIR. The as-prepared cocrystal nanofibres had smooth surfaces and uniform dimension. When incorporated into prototype devices, they exhibited good photoresponse at ambient conditions. Additionally, the phototransistor characteristics with a maximum I(on)/I(off) ratio of -460 was demonstrated. The facile synthesis and good photoresponse may boost the potential applications of cocrystal-based nanostructures in future miniaturized devices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF76259593
Cobalt, foil, 100x100mm, thickness 2.0mm, as rolled, 99.9%
Co
GF48813723
Cobalt, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Co
GF13761918
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Co
GF95441283
Cobalt, foil, 50x50mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF74432392
Cobalt, wire reel, 0.1m, diameter 0.05mm, hard, 99.9%
Co
GF20239017
Cobalt, wire reel, 1m, diameter 0.5mm, hard, 99.99+%
Co
GF41834011
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.9%
Co
GF85563535
Cobalt, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF55186050
Cobalt, wire reel, 0.2m, diameter 1.0mm, hard, 99.99+%
Co
GF12274253
Cobalt, wire reel, 0.1m, diameter 2.0mm, hard, 99.99+%
Co
GF70398502
Cobalt, wire reel, 0.2m, diameter 0.25mm, hard, 99.99+%
Co
GF29762646
Cobalt, foil, 4mm disks, thickness 0.025mm, 99.9%
Co
GF91564904
Cobalt, foil, 250x250mm, thickness 0.50mm, annealed, 99.9%
Co
GF90510094
Cobalt, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Co
GF15401135
Cobalt, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Co
GF63298198
Cobalt, wire reel, 0.05m, diameter 2.0mm, hard, 99.99+%
Co
GF54906385
Cobalt, wire reel, 0.025m, diameter 2.0mm, hard, 99.99+%
Co
GF15124081
Cobalt, wire reel, 0.1m, diameter 0.5mm, hard, 99.99+%
Co
GF60120997
Cobalt, wire reel, 0.05m, diameter 0.5mm, hard, 99.99+%
Co
GF91382069
Cobalt, wire reel, 0.1m, diameter 0.1mm, hard, 99.99+%
Co
GF94594450
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
Co
GF41541476
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Co
GF45372315
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Co
GF08560289
Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%
Co
GF89559343
Cobalt, foil, 4mm disks, thickness 0.005mm, 99.9%
Co
GF48392051
Cobalt, foil, 4mm disks, thickness 0.006mm, 99.9%
Co
GF55954873
Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Co
GF92926727
Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%
Co
GF70445246
Cobalt, foil, 4mm disks, thickness 0.009mm, 99.9%
Co
GF40089225
Cobalt, foil, 4mm disks, thickness 0.0125mm, 99.9%
Co