EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Spectroscopic properties of Yb3+ doped CaNb2O6 phosphor.
PMID 24738370
Abstract
The spectroscopic measurements of orthorhombic CaNb2O6 phosphor doped with Yb3+ ions have been performed. Low temperature spectra indicate that Yb3+ ions may occupy three types of nonequivalent sites inside the CaNb2O6 lattice: a main Yb3+ site dominating the near-infrared emission, and two other minor sites of similar behaviors to each other. An attempt to determine the Stark sublevels energies of the 2F7/2 and 2F5/2 manifolds of three Yb3+ nonequivalent ions was carried out. The fluorescence lifetimes associated to the emission lines of Yb3+ ions located at the main site and minor sites are also quite different. Furthermore, there exists energy transfer from the main site to the minor sites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248568
Calcium oxide, reagent grade
CaO
262986
Ytterbium, chips, 99.9% trace rare earth metals basis
Yb
548804
Ytterbium, chunks, 99.9% trace metals basis
Yb
261300
Ytterbium, ingot, 99.9% trace rare earth metals basis
Yb
466069
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Yb
GF49006492
Ytterbium, rod, 50mm, diameter 6.35mm, cast, 99%
Yb
GF08606516
Ytterbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Yb
GF00044682
Ytterbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Yb
GF85959444
Ytterbium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Yb
GF37805137
Ytterbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Yb
GF54792052
Ytterbium, lump, 100g, max. size 25mm, 99.9%
Yb
GF48969905
Ytterbium, lump, 25g, max. size 25mm, 99.9%
Yb
GF21238332
Ytterbium, lump, 50g, max. size 25mm, 99.9%
Yb
GF76054530
Ytterbium, rod, 100mm, diameter 10.0mm, cast, 99%
Yb
GF25811502
Ytterbium, rod, 100mm, diameter 6.35mm, cast, 99%
Yb
GF83746705
Ytterbium, rod, 50mm, diameter 10.0mm, cast, 99%
Yb
GF29517464
Ytterbium, rod, 50mm, diameter 2.0mm, cast, 99%
Yb
GF07036006
Ytterbium, wire reel, 100mm, diameter 1.0mm, square section, 99.9%
Yb
GF37587684
Ytterbium, wire reel, 300mm, diameter 1.0mm, square section, 99.9%
Yb
GF42456281
Ytterbium, wire reel, 50mm, diameter 1.0mm, square section, 99.9%
Yb
GF19264675
Ytterbium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Yb
GF49999516
Ytterbium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%
Yb
GF49939655
Ytterbium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Yb