EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Application of solution-processed V2O5 in inverted polymer solar cells based on fluorine-doped tin oxide substrate.
PMID 24738373
Abstract
We used a hydrothermal method to synthesis the solution-processed V2O5 as anode buffer layer, which applied on inverted polymer solar cells based on FTO substrate. The structure of the device is glass/FTO/TiO2/P3HT:PCBM/V2O5/Ag. We discussed the dependence of device performance on the concentrations of V2O5 solution. It is found that when the concentration of V2O5 is 300 microg/ml, the power conversion efficiency (PCE of 2.38%) is the highest, which is much higher than that of the device without anode buffer layer (PCE of only 0.87%). Moreover, it can significantly reduce the energy consumption and make it more cost-effective.