EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Pr3+/Yb3+ co-doped beta-phase NaYF4 microprisms: controlled synthesis and upconversion luminescence.
PMID 24738388
Abstract
Pr3+/Yb3+ co-doped hexagonal NaYF4(beta-NaYF4) microprisms were synthesized by the hydrothermal method, and ethylenediaminetetraacetic acid (EDTA) was introduced to control the size of the microcrystal samples. Bright upconverted fluorescence emission was observed when the samples were excited with an infrared (IR) laser at 976.4 nm. The emission was found to originate from the transitions of 3P0-3F2, 3P0-3H6 or 1G4-3H4, 3P1-3H6, 3P0-3H5, 3P1-3H5, and 3P0-3H4 of Pr3+ ions. Possible mechanisms for upconversion fluorescence and concentration dependence as well as the crystal structure and its formation of NaYF4:Yb3+/Pr3+ microprisms were explored and discussed based on the experimental observations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261173
Praseodymium, ingot, under oil, 99.9% trace rare earth metals basis
Pr
263176
Praseodymium, powder, ~40 mesh, 99.5% trace rare earth metals basis
Pr
GF48004804
Praseodymium, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99%
Pr
GF66110329
Praseodymium, rod, 100mm, diameter 6.35mm, cast, 99%
Pr
GF06128883
Praseodymium, rod, 50mm, diameter 6.35mm, cast, 99%
Pr
GF50132191
Praseodymium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Pr
GF66524643
Praseodymium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Pr
GF92630246
Praseodymium, foil, not light tested, 50x50mm, thickness 0.015mm, as rolled, 99%
Pr
GF34755857
Praseodymium, foil, not light tested, 50x50mm, thickness 0.005mm, as rolled, 99%
Pr
GF82608295
Praseodymium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Pr
GF69747621
Praseodymium, foil, not light tested, 25x25mm, thickness 0.01mm, as rolled, 99%
Pr
GF75299170
Praseodymium, foil, not light tested, 25x25mm, thickness 0.015mm, as rolled, 99%
Pr
GF96026531
Praseodymium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Pr
GF07601059
Praseodymium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Pr
GF91191191
Praseodymium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%
Pr
GF98899687
Praseodymium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Pr
262986
Ytterbium, chips, 99.9% trace rare earth metals basis
Yb
548804
Ytterbium, chunks, 99.9% trace metals basis
Yb
261300
Ytterbium, ingot, 99.9% trace rare earth metals basis
Yb
466069
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Yb
GF49006492
Ytterbium, rod, 50mm, diameter 6.35mm, cast, 99%
Yb
GF08606516
Ytterbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Yb
GF00044682
Ytterbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Yb
GF85959444
Ytterbium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Yb
GF37805137
Ytterbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Yb
GF54792052
Ytterbium, lump, 100g, max. size 25mm, 99.9%
Yb
GF48969905
Ytterbium, lump, 25g, max. size 25mm, 99.9%
Yb
GF21238332
Ytterbium, lump, 50g, max. size 25mm, 99.9%
Yb
GF76054530
Ytterbium, rod, 100mm, diameter 10.0mm, cast, 99%
Yb
GF25811502
Ytterbium, rod, 100mm, diameter 6.35mm, cast, 99%
Yb