EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
The effect of growth temperature variation on partially bismuth filled carbon nanotubes synthesis using a soft semi-metallic template.
PMID 24738435
Abstract
The dewetting of a low melting point metal thin film deposited on silicon substrates was studied. The experimental results suggest that the change in the growth temperature affects the nanostructures that form. Based on the experimental results, the temperature which yielded the smallest features for the growth of nanotubes is determined. The mechanism by which these nano-templates become an efficient seeds for the growth of the carbon nanotubes is discussed. The partial bismuth filling inside the CNTs was optimized. Based on the results, a schematic growth model for better understanding of the process parameters has also been proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF86561354
Bismuth, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF85045631
Bismuth, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF80139438
Bismuth, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF60966366
Bismuth, rod, 70mm, diameter 10.0mm, cast sticks, 100%
Bi
GF89918500
Bismuth, foil, 25x25mm, thickness 0.25mm, 100%
Bi
GF71570550
Bismuth, foil, 25x25mm, thickness 0.5mm, 100%
Bi
GF58555581
Bismuth, foil, 25x25mm, thickness 1.0mm, 100%
Bi
GF52383506
Bismuth, foil, 25x25mm, thickness 1.0mm, 99.97%
Bi
GF31001960
Bismuth, foil, 25x25mm, thickness 2.0mm, 100%
Bi
GF39813419
Bismuth, foil, 50x50mm, thickness 0.25mm, 100%
Bi
GF46319340
Bismuth, foil, 50x50mm, thickness 0.5mm, 100%
Bi
GF19448017
Bismuth, foil, 50x50mm, thickness 1.0mm, 100%
Bi
GF97656238
Bismuth, foil, 50x50mm, thickness 1.0mm, 99.97%
Bi
GF75246183
Bismuth, foil, 50x50mm, thickness 2.0mm, 100%
Bi
GF46489135
Bismuth, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF86892441
Bismuth, foil, light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF40931546
Bismuth, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF35246831
Bismuth, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF95571995
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF12550935
Bismuth, foil, light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF09655263
Bismuth, foil, light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF71673060
Bismuth, foil, light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF09816307
Bismuth, foil, light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF91742328
Bismuth, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF24083692
Bismuth, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 99.97%
Bi
GF77293592
Bismuth, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 99.97%
Bi
GF93912891
Bismuth, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi
GF19216087
Bismuth, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.97%
Bi
GF64303455
Bismuth, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.97%
Bi
GF30816356
Bismuth, foil, not light tested, 150x150mm, thickness 0.025mm, permanent polyester support, 99.97%
Bi