EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
Design, characterization, and aerosol dispersion performance modeling of advanced co-spray dried antibiotics with mannitol as respirable microparticles/nanoparticles for targeted pulmonary delivery as dry powder inhalers.
PMID 24740732