EMAIL THIS PAGE TO A FRIEND
Journal of neurophysiology
A potentially novel nicotinic receptor in Aplysia neuroendocrine cells.
PMID 24740855
Abstract
Nicotinic receptors form a diverse group of ligand-gated ionotropic receptors with roles in both synaptic transmission and the control of excitability. In the bag cell neurons of Aplysia, acetylcholine activates an ionotropic receptor, which passes inward current to produce a long-lasting afterdischarge and hormone release, leading to reproduction. While testing the agonist profile of the cholinergic response, we observed a second current that appeared to be gated only by nicotine and not acetylcholine. The peak nicotine-evoked current was markedly smaller in magnitude than the acetylcholine-induced current, cooperative (Hill value of 2.7), had an EC50 near 500 μM, readily recovered from desensitization, showed Ca(2+) permeability, and was blocked by mecamylamine, dihydro-β-erythroidine, or strychnine, but not by α-conotoxin ImI, methyllycaconitine, or hexamethonium. Aplysia transcriptome analysis followed by PCR yielded 20 full-length potential nicotinic receptor subunits. Sixteen of these were predicted to be cation selective, and real-time PCR suggested that 15 of the 16 subunits were expressed to varying degrees in the bag cell neurons. The acetylcholine-induced current, but not the nicotine current, was reduced by double-strand RNA treatment targeted to both subunits ApAChR-C and -E. Conversely, the nicotine-evoked current, but not the acetylcholine current, was lessened by targeting both subunits ApAChR-H and -P. To the best of our knowledge, this is the first report suggesting that a nicotinic receptor is not gated by acetylcholine. Separate receptors may serve as a means to differentially trigger plasticity or safeguard propagation by assuring that only acetylcholine, the endogenous agonist, initiates large enough responses to trigger reproduction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36733
(−)-Nicotine, PESTANAL®, analytical standard
C10H14N2
D5891
1,1-Dimethyl-4-phenylpiperazinium iodide, ≥98% (TLC or titration), powder
C12H19IN2
A2661
Acetylcholine chloride, suitable for cell culture
C7H16ClNO2
A9101
Acetylcholine chloride, pkg of 150 mg (per vial)
C7H16ClNO2
1008501
Acetylcholine chloride, United States Pharmacopeia (USP) Reference Standard
C7H16ClNO2
Y0000002
Acetylcholine chloride, European Pharmacopoeia (EP) Reference Standard
C7H16ClNO2
A3377
Adenosine 5′-triphosphate disodium salt hydrate, Grade II, ≥98.5% (HPLC), crystalline, from microbial
C10H14N5Na2O13P3 · xH2O
G6752
Guanosine, ≥98%
C10H13N5O5
G6264
Guanosine, suitable for cell culture, BioReagent
C10H13N5O5
75903
Guanosine, ≥97.0% (HPLC)
C10H13N5O5
G7637
Guanosine 5′-[β-thio]diphosphate trilithium salt, ≥85% (HPLC), powder
C10H12Li3N5O10P2S
G8877
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
C10H16N5O14P3 · xNa+
H0879
Hexamethonium bromide
C12H30Br2N2
H2138 Hexamethonium chloride, solid (hygroscopic)
C12H30Cl2N2
438065
Hydrobromic acid, ACS reagent, 48%
HBr
18735
Hydrobromic acid solution, 33 wt. % in acetic acid
HBr
295418
Hydrogen bromide, ReagentPlus®, ≥99%
HBr
248630
Hydrogen bromide solution, 33 wt. % in acetic acid
HBr
M5255
Mycophenolic acid, ≥98%
C17H20O6
M3536
Mycophenolic acid, powder, BioReagent, suitable for cell culture
C17H20O6
89287
Mycophenolic acid, analytical standard
C17H20O6