EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Gastrointestinal and Liver Physiology
Acute murine colitis reduces colonic 5-aminosalicylic acid metabolism by regulation of N-acetyltransferase-2.
PMID 24742986