EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Successful interferon-alpha 2b therapy for unremitting warts in a patient with DOCK8 deficiency.
PMID 24743019
Abstract
The autosomal recessive form of the Hyper IgE syndrome (AR-HIES) with dedicator of cytokinesis 8 (DOCK8) deficiency is associated with difficult to treat persistent viral skin infections, including papilloma virus infection. Type I interferons play an important role in the defense against viruses. We examined the effect of therapy with IFN-α 2b in an 11-year old boy with DOCK8 deficiency due to a homozygous splice donor site mutation in DOCK8 intron 40. His unremitting warts showed dramatic response to IFN-α 2b therapy. Immunological studies revealed decreased circulating plasmacytoid dendritic cells (pDCs) and profound deficiency of IFN-α production by his peripheral blood mononuclear cells in response to treatment with CpG oligonucleotides. These findings indicate that underlying pDC deficiency and impaired IFN-α production may predispose to chronic viral infections in DOCK8 deficiency. IFN-α 2b therapy maybe useful in controlling recalcitrant viral infections in these patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0320301 Interferon α-2b, European Pharmacopoeia (EP) Reference Standard