EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Pseudomonas hussainii sp. nov., isolated from droppings of a seashore bird, and emended descriptions of Pseudomonas pohangensis, Pseudomonas benzenivorans and Pseudomonas segetis.
PMID 24744016
Abstract
Two Gram-staining-negative, aerobic, rod-shaped, non-spore-forming bacterial strains that are motile by a monopolar flagellum, designated CC-AMH-11(T) and CC-AMHZ-5, were isolated from droppings of a seashore bird off the coast of Hualien, Taiwan. The strains showed 99.7% mutual pairwise 16S rRNA gene sequence similarity, while exhibiting <96.2% sequence similarity to strains of other species of the genus Pseudomonas (95.7-95.9% similarity with type species, Pseudomonas aeruginosa LMG 1242T), and formed a distinct co-phyletic lineage in the phylogenetic trees. The common major fatty acids (>5% of the total) were C18 : 1ω7c and/or C18 : 1ω6c (summed feature 8), C16 : 1ω6c and/or C16 : 1ω7c (summed feature 3), C16 : 0 and C12 : 0. Phosphatidylethanolamine, phosphatidylglycerol, diphosphatidylglycerol, phosphatidylserine, an unidentified lipid and an unidentified phospholipid were detected as common polar lipids. The DNA G+C contents of strains CC-AMH-11(T) and CC-AMHZ-5 were 61.1 and 61.6 mol%, respectively. The common major respiratory quinone was ubiquinone 9 (Q-9), and the predominant polyamine was putrescine. The DNA-DNA hybridization obtained between the two strains was 79.0% (reciprocal value 89.4% using CC-AMHZ-5 DNA as the probe). The very high 16S rRNA gene sequence similarity and DNA-DNA relatedness and the poorly distinguishable phenotypic features witnessed between CC-AMH-11(T) and CC-AMHZ-5 suggested unambiguously that they are two distinct strains of a single genomic species. However, the strains also showed several genotypic and phenotypic characteristics that distinguished them from other closely related species of Pseudomonas. Thus, the strains are proposed to represent a novel species of Pseudomonas, for which the name Pseudomonas hussainii sp. nov. is proposed. The type strain is CC-AMH-11(T) ( = JCM 19513(T) = BCRC 80696(T)); a second strain of the same species is CC-AMHZ-5 ( = JCM 19512 = BCRC 80697). In addition, emended descriptions of the species Pseudomonas pohangensis, Pseudomonas benzenivorans and Pseudomonas segetis are also proposed.