EMAIL THIS PAGE TO A FRIEND
Human pathology
Aberrant expression of p53, p21, cyclin D1, and Bcl2 and their clinicopathological correlation in ampullary adenocarcinoma.
PMID 24746206
Abstract
Previous studies on the molecular alterations in ampullary adenocarcinoma (AA) are limited, and little is known about their clinical implications. The objective of this study is to examine the expression of p53, p21, cyclin D1, and Bcl2 and their clinical significance in patients with AA. Tissue microarrays were constructed using archival tissue from 92 patients with AA who underwent pancreaticoduodenectomy at our institution. Each tumor was sampled in triplicate with a 1.0-mm punch from representative areas. The expression of p53, p21, cyclin D1, and Bcl2 was evaluated by immunohistochemistry, and the staining results were correlated with clinicopathological features and survival. Among 92 cases studied, overexpression of p53, p21, cyclin D1, and Bcl2 was observed in 58.7%, 39.2%, 71.7%, and 5.4% of tumors, respectively. Patients whose tumor showed high level of cyclin D1 expression had higher risk of disease recurrence (P = .02) and worse recurrence-free and overall survivals after pancreaticoduodenectomy than did those with no or low cyclin D1 expression (P = .027 and P = .02, respectively). In multivariate analysis, cyclin D1 expression was an independent prognostic factor for both recurrence-free and overall survival (P < .05). However, there was no significant correlation between p53, p21, or Bcl2 expression and survival (P > .05). Our study showed that p53, p21, and cyclin D1, but not Bcl2, are frequently overexpressed in AAs. Cyclin D1 overexpression is associated with increased risk of disease recurrence and worse survival in patients with AA after resection.
Related Materials