EMAIL THIS PAGE TO A FRIEND
Organic letters
Trisubstituted 2-trifluoromethyl pyrrolidines via catalytic asymmetric Michael addition/reductive cyclization.
PMID 24746344
Abstract
The stereoselective synthesis of trisubstituted 2-trifluoromethyl pyrrolidines by asymmetric Michael addition/hydrogenative cyclization is described. The direct organocatalytic addition of 1,1,1-trifluoromethylketones to nitroolefins proceeds under mild reaction conditions and low catalyst loadings to provide Michael adducts in high yield with excellent diastereo- and enantioselectivity. Catalytic hydrogenation of the Michael adducts stereoselectively generates 2-trifluoromethylated pyrrolidines bearing three contiguous stereocenters. A stereospecific route to epimeric 2-trifluoromethyl pyrrolidines from a common intermediate is described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
803863
Schreiner′s Thiourea Catalyst, 95%
C17H8F12N2S