EMAIL THIS PAGE TO A FRIEND
Bioresource Technology
Boosting accumulation of neutral lipids in Rhodosporidium kratochvilovae HIMPA1 grown on hemp (Cannabis sativa Linn) seed aqueous extract as feedstock for biodiesel production.
PMID 24746767
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5252
Nonadecanoic acid, ≥98% (GC)
C19H38O2
72332
Nonadecanoic acid, analytical standard
C19H38O2