EMAIL THIS PAGE TO A FRIEND
Brain Structure &Amp; Function
Pre-existing astrocytes form functional perisynaptic processes on neurons generated in the adult hippocampus.
PMID 24748560
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB377
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse