EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
PCL/PEG core/sheath fibers with controlled drug release rate fabricated on the basis of a novel combined technique.
PMID 24751343
Abstract
A novel core/sheath fiber preparation method, which included the processes of blend electrospinning to produce the core fiber and UV-induced graft polymerization to fabricate the outer polymeric shell, was presented to provide designated fibers with different shell thicknesses. A hydrophilic drug, salicylic acid (SA), was loaded in the representative poly(ϵ-caprolactone) (PCL)/polyethylene glycol (PEG) core/sheath fibers, performed according to this combined technique. FTIR analysis indicated that the existence of hydrogen bonds between SA and the PCL matrix improved drug compatibility. Field emission scanning electron microscopy (FESEM) images indicated that the morphology and the diameter distribution of fibers changed significantly after the graft polymerization procedure. All the core/sheath fibers became more flexible and thicker compared with the core fiber. The water contact angle (WCA) test also noted the differences of these two fibers: PCL/PEG core/sheath fibers with cross-linked PEG surface exhibited more hydrophilic property. Moreover, in vitro SA release tests were conducted to explore the relationship between the PEG shell thickness and the drug release rate. A typical biphasic release mechanism was observed for the PCL/PEG core/sheath fibers, and their sustained release rates were controlled by the PEG shell thickness in a linear correlation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
427551
Benzophenone, purified by sublimation, ≥99%
C13H10O
B9300
Benzophenone, ReagentPlus®, 99%
C13H10O
W213403
Benzophenone, ≥99%, FCC, FG
C13H10O
12750
Benzophenone, purum, ≥99.0% (GC)
C13H10O
442842
Benzophenone, analytical standard
C13H10O
Y0000647
Benzophenone, European Pharmacopoeia (EP) Reference Standard
C13H10O
PHR1204
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
C13H10O
50296
Melting point standard 47-49°C, analytical standard
C13H10O
73664
Mettler-Toledo Calibration substance ME 18870, Benzophenone, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C13H10O
S5922
Salicylic acid, BioXtra, ≥99.0%
C7H6O3
S7401
Salicylic acid, plant cell culture tested
C7H6O3
247588
Salicylic acid, ACS reagent, ≥99.0%
C7H6O3
84210
Salicylic acid, puriss. p.a., ≥99.0% (T)
C7H6O3
27301
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
C7H6O3
1609002
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O3
84211
Salicylic acid, tested according to Ph.Eur.
C7H6O3
PHR1013
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O3
105910
Salicylic acid, ReagentPlus®, ≥99%
C7H6O3
W398500
Salicylic acid, ≥99%, FG
C7H6O3
52341
Salicylic acid, certified reference material, TraceCERT®
C7H6O3
S0200000
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O3
S-019
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material
C7H6O3