EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Screening of specific inhibitors for human carboxylesterases or arylacetamide deacetylase.
PMID 24751575
Abstract
Esterases catalyze the hydrolysis of therapeutic drugs containing esters or amides in their structures. Human carboxylesterase (CES) and arylacetamide deacetylase (AADAC) are the major enzymes that catalyze the hydrolysis of drugs in the liver. Characterization of the enzyme(s) responsible for drug metabolism is required in drug development and to realize optimal drug therapy. Because multiple enzymes may show a metabolic potency for a given compound, inhibition studies using chemical inhibitors are useful tools to determine the contribution of each enzyme in human tissue preparations. The purpose of this study was to find specific inhibitors for human CES1, CES2, and AADAC. We screened 542 chemicals for the inhibition potency toward hydrolase activities of p-nitrophenyl acetate by recombinant CES1, CES2, and AADAC. We found that digitonin and telmisartan specifically inhibited CES1 and CES2 enzyme activity, respectively. Vinblastine potently inhibited both AADAC and CES2, but no specific inhibitor of AADAC was found. The inhibitory potency and specificity of these compounds were also evaluated by monitoring the effects on hydrolase activity of probe compounds of each enzyme (CES1: lidocaine, CES2: CPT-11, AADAC: phenacetin) in human liver microsomes. Telmisartan and vinblastine strongly inhibited the hydrolysis of CPT-11 and/or phenacetin, but digitonin did not strongly inhibit the hydrolysis of lidocaine, indicating that the inhibitory potency of digitonin was different between recombinant CES1 and liver microsomes. Although we could not find a specific inhibitor of AADAC, the combined use of telmisartan and vinblastine could predict the responsibility of human AADAC to drug hydrolysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS008318
(4S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3′,4′:6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)dione, AldrichCPR
C22H20N2O5
D146005
2,6-Dimethylaniline, 99%
C8H11N
36766
2,6-Dimethylaniline, PESTANAL®, analytical standard
C8H11N
39520
2,6-Dimethylaniline, purum, ≥98.0% (GC)
C8H11N
442327
2,6-Dimethylaniline, analytical standard
C8H11N
P14815
4-Ethoxyaniline, 98%
C8H11NO
1048
4-Nitrophenol, spectrophotometric grade
C6H5NO3
35836
4-Nitrophenol, PESTANAL®, analytical standard
C6H5NO3
48549
4-Nitrophenol
C6H5NO3
N7660
4-Nitrophenol solution, 10 mM
C6H5NO3
43575
4-Nitrophenol solution, stock solution, for mass spectrometry
C6H5NO3
40056
4-Nitrophenol solution, certified reference material, 5000 μg/mL in methanol
C6H5NO3
49053
4-Nitrophenol solution, working solution, for mass spectrometry
C6H5NO3
N8130
4-Nitrophenyl acetate, esterase substrate
C8H7NO4
46021
4-Nitrophenyl acetate, ≥99.0% (GC)
C8H7NO4
H0165
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
C22H20N2O5
PHR1150
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
C6H5NO3
Y0001575
Bupivacaine impurity F, European Pharmacopoeia (EP) Reference Standard
C8H11N
I1406
Irinotecan hydrochloride, topoisomerase inhibitor
C33H38N4O6 · HCl
77440
Phenacetin, ≥98.0% (HPLC)
C10H13NO2
PHR1249
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
C10H13NO2
PHR1094
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C10H13NO2
1514008
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C10H13NO2