EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Calorimetric investigation of diclofenac drug binding to a panel of moderately glycated serum albumins.
PMID 24751671
Abstract
Glycation alters the drug binding properties of serum proteins and could affect free drug concentrations in diabetic patients with elevated glycation levels. We investigated the effect of bovine serum albumin glycation by eight physiologically relevant glycation reagents (glucose, ribose, carboxymethyllysine, acetoin, methylglyoxal, glyceraldehyde, diacetyl and glycolaldehyde) on diclofenac drug binding. We used this non-steroidal anti-inflammatory drug diclofenac as a paradigm for acidic drugs with high serum binding and because of its potential cardiovascular risks in diabetic patients. Isothermal titration calorimetry showed that glycation reduced the binding affinity Ka of serum albumin and diclofenac 2 to 6-fold by reducing structural rigidity of albumin. Glycation affected the number of drug binding sites in a glycation reagent dependent manner and lead to a 25% decrease for most reagent, expect for ribose, with decreased by 60% and for the CML-modification, increased the number of binding sites by 60%. Using isothermal titration calorimetry and differential scanning calorimetry we derived the complete thermodynamic characterization of diclofenac binding to all glycated BSA samples. Our results suggest that glycation in diabetic patients could significantly alter the pharmacokinetics of the widely used over-the-counter NSDAI drug diclofenac and with possibly negative implications for patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
260150
Glyoxylic acid solution, 50 wt. % in H2O
C2H2O3