EMAIL THIS PAGE TO A FRIEND
Developmental dynamics : an official publication of the American Association of Anatomists
Role of heparan sulfate proteoglycans in optic disc and stalk morphogenesis.
PMID 24753163
Abstract
Heparan sulfate proteoglycans (HSPG) are important for embryonic development by means of the regulation of gradient formation and signaling of multiple growth factors and morphogens. Previous studies have shown that Bmp/Shh/Fgf signaling are required for the regionalization of the optic vesicle (OV) and for the closure of the optic fissure (OF), the disturbance of which underlie ocular anomalies such as microphthalmia, coloboma, and optic nerve hypoplasia. To study HSPG-dependent coordination of these signaling pathways during mammalian visual system development, we have generated a series of OV-specific mutations in the heparan sulfate (HS) N-sulfotransferase genes (Ndst1 and Ndst2) and HS O-sulfotransferase genes (Hs2st, Hs6st1, and Hs6st2) in mice. Of interest, the resulting HS undersulfation still allowed for normal retinal neurogenesis and optic fissure closure, but led to defective optic disc and stalk development. The adult mutant animals further developed optic nerve aplasia/hypoplasia and displayed retinal degeneration. We observed that MAPK/ERK signaling was down-regulated in Ndst mutants, and consistent with this, HS-related optic nerve morphogenesis defects in mutant mice could partially be rescued by constitutive Kras activation. These results suggest that HSPGs, depending on their HS sulfation pattern, regulate multiple signaling pathways in optic disc and stalk morphogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100362
Anti-CCND1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100363
Anti-CCND1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2100364
Anti-CCND1 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB1405510
Anti-CCND1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
GW21421 Anti-MITF antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA003259
Anti-MITF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105416
Anti-MITF antibody produced in rabbit, affinity isolated antibody
SAB4501879
Anti-MITF antibody produced in rabbit, affinity isolated antibody
SAB2501318
Anti-NEFM antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA022845
Anti-NEFM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA023138
Anti-NEFM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA047704
Anti-PAX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410876
Anti-PAX2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300194
Anti-phospho-MAPK3 (pThr202) antibody produced in rabbit, affinity isolated antibody
SAB4300195
Anti-phospho-MAPK3 (pTyr204) antibody produced in rabbit, affinity isolated antibody
SAB4504626
Anti-phospho-MNK1 (pThr255) antibody produced in rabbit, affinity isolated antibody
SAB4504714
Anti-phospho-Mnk1 (pThr385) antibody produced in rabbit, affinity isolated antibody
AB5603
Anti-Sox2 Antibody, Anti-SOX2 Antibody, Cat. No. AB5603, is a highly specific rabbit polyclonal antibody SOX2 and has been tested for use in Immunocytochemistry, and Immunohistochemistry (Paraffin), and Western Blotting.
SAB2701800
Anti-sox2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S9072 Anti-SOX2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500363
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300669
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB2701973
Anti-SOX2 antibody produced in rabbit
SAB2701974
Anti-SOX2 antibody produced in rabbit
SAB2103958
Anti-SOX2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SRP3262 FGF-10 human, Animal-component free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
SAB4700017 Monoclonal Anti-CCND1 antibody produced in mouse, clone CD1.1, purified immunoglobulin, buffered aqueous solution
C7464
Monoclonal Anti-Cyclin D1 antibody produced in mouse, clone DCS-6, ascites fluid
WH0005076M1
Monoclonal Anti-PAX2 antibody produced in mouse, clone 3C7, purified immunoglobulin, buffered aqueous solution
S1451
Monoclonal Anti-Sox2 antibody produced in mouse, ~1.0 mg/mL, clone SOX2-6, purified immunoglobulin, buffered aqueous solution