EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[A new phthalide from angelicae sinensis radix].
PMID 24754173
Abstract
Angelicae Sinensis Radix, dried roots of Angelicae sinensis, is a well-known traditional Chinese medicine. Six compounds were isolated and purified by silica gel and preparative HPLC. According to physicochemical properties and spectral data, the compounds were identified as senkyunolide H-7-acetate (1), o-phthalic acid (2), diisobutyl phthalate (3), p-hydroxyphenylethanol ferulate (4), ferulic acid (5) and coniferylferulate (6). Compound 1 was a new one.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90034
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical standard
C10H10O4