EMAIL THIS PAGE TO A FRIEND
Brain : a journal of neurology
Pharmacological stimulation of sigma-1 receptors has neurorestorative effects in experimental parkinsonism.
PMID 24755275
Abstract
The sigma-1 receptor, an endoplasmic reticulum-associated molecular chaperone, is attracting great interest as a potential target for neuroprotective treatments. We provide the first evidence that pharmacological modulation of this protein produces functional neurorestoration in experimental parkinsonism. Mice with intrastriatal 6-hydroxydopamine lesions were treated daily with the selective sigma-1 receptor agonist, PRE-084, for 5 weeks. At the dose of 0.3 mg/kg/day, PRE-084 produced a gradual and significant improvement of spontaneous forelimb use. The behavioural recovery was paralleled by an increased density of dopaminergic fibres in the most denervated striatal regions, by a modest recovery of dopamine levels, and by an upregulation of neurotrophic factors (BDNF and GDNF) and their downstream effector pathways (extracellular signal regulated kinases 1/2 and Akt). No treatment-induced behavioural-histological restoration occurred in sigma-1 receptor knockout mice subjected to 6-hydroxydopamine lesions and treated with PRE-084. Immunoreactivity for the sigma-1 receptor protein was evident in both astrocytes and neurons in the substantia nigra and the striatum, and its intracellular distribution was modulated by PRE-084 (the treatment resulted in a wider intracellular distribution of the protein). Our results suggest that sigma-1 receptor regulates endogenous defence and plasticity mechanisms in experimental parkinsonism. Boosting the activity of this protein may have disease-modifying effects in Parkinson's disease.