EMAIL THIS PAGE TO A FRIEND
Cancer Biology & Therapy
Etoposide-loaded immunoliposomes as active targeting agents for GD2-positive malignancies.
PMID 24755919
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0503
O-Phosphorylethanolamine
C2H8NO4P