EMAIL THIS PAGE TO A FRIEND
JAMA
Easy-to-use overdose antidote earns fast-track approval.
PMID 24756492
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000695
Naloxone for peak identification, European Pharmacopoeia (EP) Reference Standard
C19H21NO4 · HCl · 2H2O