EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
Identification of cadherin 11 as a mediator of dermal fibrosis and possible role in systemic sclerosis.
PMID 24757152
Abstract
Systemic sclerosis (SSc) is a chronic autoimmune disease clinically manifesting as progressive fibrosis of the skin and internal organs. Recent microarray studies demonstrated that cadherin 11 (Cad-11) expression is increased in the affected skin of patients with SSc. The purpose of this study was to examine our hypothesis that Cad-11 is a mediator of dermal fibrosis. Biopsy samples of skin from SSc patients and healthy control subjects were used for real-time quantitative polymerase chain reaction analysis to assess Cad-11 expression and for immunohistochemistry to determine the expression pattern of Cad-11. To determine whether Cad-11 is a mediator of dermal fibrosis, Cad-11-deficient mice and anti-Cad-11 monoclonal antibodies (mAb) were used in the bleomycin-induced dermal fibrosis model. In vitro studies with dermal fibroblasts and bone marrow-derived macrophages were used to determine the mechanisms by which Cad-11 contributes to the development of tissue fibrosis. Levels of messenger RNA for Cad-11 were increased in skin biopsy samples from patients with SSc and correlated with the modified Rodnan skin thickness scores. Cad-11 expression was localized to dermal fibroblasts and macrophages in SSc skin. Cad-11-knockout mice injected with bleomycin had markedly attenuated dermal fibrosis, as quantified by measurements of skin thickness, collagen levels, myofibroblast accumulation, and profibrotic gene expression, in lesional skin as compared to the skin of wild-type mice. In addition, anti-Cad-11 mAb decreased fibrosis at various time points in the bleomycin-induced dermal fibrosis model. In vitro studies demonstrated that Cad-11 regulated the production of transforming growth factor β (TGFβ) by macrophages and the migration of fibroblasts. These data demonstrate that Cad-11 is a mediator of dermal fibrosis and TGFβ production and suggest that Cad-11 may be a therapeutic target in SSc.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300563
Anti-SMAD2 (Ab-220) antibody produced in rabbit, affinity isolated antibody
SAB4300562
Anti-SMAD2 (Ab-467) antibody produced in rabbit, affinity isolated antibody
SAB4200354
Anti-SMAD2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4501805
Anti-SMAD2 antibody produced in rabbit, affinity isolated antibody
SAB4501806
Anti-SMAD2 antibody produced in rabbit, affinity isolated antibody
SAB4501807
Anti-SMAD2 antibody produced in rabbit, affinity isolated antibody
SAB4501808
Anti-SMAD2 antibody produced in rabbit, affinity isolated antibody
SAB2103328
Anti-SMAD2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SRP3211 IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
I1020 Interleukin-4 from mouse, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
SAB4100238
Monoclonal Anti-SMAD2 antibody produced in mouse, clone SMAD2-B203, culture supernatant
SAB1404036
Monoclonal Anti-SMAD2 antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution
SAB1400158
Monoclonal Anti-SMAD2 antibody produced in mouse, clone 3G6, purified immunoglobulin, buffered aqueous solution
SAB1400159
Monoclonal Anti-SMAD2 antibody produced in mouse, clone 3G9, purified immunoglobulin, buffered aqueous solution
WH0004087M1
Monoclonal Anti-SMAD2 antibody produced in mouse, clone 4D10, purified immunoglobulin, buffered aqueous solution
AV38694
Anti-SMAD2 antibody produced in rabbit, IgG fraction of antiserum