EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Henna tattoo: infection or allergy?
PMID 24759251
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H46805
2-Hydroxy-1,4-naphthoquinone, 97%
C10H6O3