EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
CarbAcineto NP test for rapid detection of carbapenemase-producing Acinetobacter spp.
PMID 24759709
Abstract
Multidrug-resistant Acinetobacter baumannii isolates, particularly those that produce carbapenemases, are increasingly reported worldwide. The biochemically based Carba NP test, extensively validated for the detection of carbapenemase producers among Enterobacteriaceae and Pseudomonas spp., has been modified to detect carbapenemase production in Acinetobacter spp. A collection of 151 carbapenemase-producing and 69 non-carbapenemase-producing Acinetobacter spp. were tested using the Carba NP test and a modified Carba NP protocol (the CarbAcineto NP test) in this study. The CarbAcineto NP test requires modified lysis conditions and an increased bacterial inoculum compared to those of the original Carba NP test. The Carba NP test detects metallo-β-lactamase producers but failed to detect the production of other carbapenemase types among Acinetobacter spp. In contrast, the newly designed CarbAcineto NP test, which is rapid and reproducible, detects all types of carbapenemases with a sensitivity of 94.7% and a specificity of 100%. This cost-effective technique offers a reliable and affordable technique for identifying carbapenemase production in Acinetobacter spp., which is a marker of multidrug resistance in those species. Its use will facilitate the recognition of these carbapenemases and prevent their spread.