EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
LMTK3 deficiency causes pronounced locomotor hyperactivity and impairs endocytic trafficking.
PMID 24760852
Abstract
LMTK3 belongs to the LMTK family of protein kinases that are predominantly expressed in the brain. Physiological functions of LMTK3 and other members of the LMTK family in the CNS remain unknown. In this study, we performed a battery of behavioral analyses using Lmtk3(-/-) mice and showed that these mice exhibit abnormal behaviors, including pronounced locomotor hyperactivity, reduced anxiety behavior, and decreased depression-like behavior. Concurrently, the dopamine metabolite levels and dopamine turnover rate are increased in the striata of Lmtk3(-/-) mice compared with wild-type controls. In addition, using cultured primary neurons from Lmtk3(-/-) mice, we found that LMTK3 is involved in the endocytic trafficking of N-methyl-d-aspartate receptors, a type of ionotropic glutamate receptor. Altered membrane traffic of the receptor in Lmtk3(-/-) neurons may underlie behavioral abnormalities in the mutant animals. Together, our data suggest that LMTK3 plays an important role in regulating locomotor behavior in mice.