EMAIL THIS PAGE TO A FRIEND
Journal of Microencapsulation
Skin penetration and photoprotection of topical formulations containing benzophenone-3 solid lipid microparticles prepared by the solvent-free spray-congealing technique.
PMID 24766207