EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Development of a multianalyte method based on micro-matrix-solid-phase dispersion for the analysis of fragrance allergens and preservatives in personal care products.
PMID 24767833