EMAIL THIS PAGE TO A FRIEND
The Journal of thoracic and cardiovascular surgery
Use of polytetrafluoroethylene vascular graft to cover the kinking protector of left ventricular assist device facilitates later pump exchange.
PMID 24768102
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
665800
Polytetrafluoroethylene preparation, 60 wt % dispersion in H2O