EMAIL THIS PAGE TO A FRIEND
Acta physiologica Scandinavica
Increased levels of substance P and cholecystokinin in rat cerebral cortex following repeated electroconvulsive shock and subchronic treatment with a serotonin uptake inhibitor.
PMID 2476913