Description
Molecular Formula
Add to Cart
N7271 α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
N5521 α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
N3786
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
N8271 α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
N5254 Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
N2876
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
N3001
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-10 units/mg protein (using 4MU-NANA), 2-5 units/mg protein (mucin)
N5631
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
N2133
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
N7885
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
N6514
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
72197
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥1.5 U/mg protein