EMAIL THIS PAGE TO A FRIEND
Voprosy virusologii
[Evaluation of the anti-neuraminidase antibodies in clinical trials of the live influenza vaccine of the A(H5N2) subtype].
PMID 24772644
Abstract
In the current study, we evaluated the neuraminidase-inhibition (NI) antibodies among volunteers during the phase I and phase II of the clinical trials of a monovalent live attenuated influenza vaccine (LAIV) A/17/duck/ Potsdam/86/92(H5N2). The reassortant influenza virus RN2/57-human A(H7N2) containing neuraminidase (NA) from the A/Leningrad/134/17/57(H2N2) was used in NI test. It was shown that two doses of the monovalent LAIV A(H5N2) led to a statistically significant increase in the NI antibodies to vaccine strain NA. More than twofold increase in antibodies was obtained among 19.5-33.3% of vaccinated. The microneutralization test and NI assay results coincidence in the same pairs of sera of the vaccinated volunteers was 73.2%, suggesting thus a statistically significant interdependence between the values of increase in antibodies revealed in both tests (p = 0.04).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7271 α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
N5521 α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
N3786
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
N8271 α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
N5254 Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
N2876
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
N3001
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-10 units/mg protein (using 4MU-NANA), 2-5 units/mg protein (mucin)
N5631
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
N2133
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
N7885
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
N6514
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
72197
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥1.5 U/mg protein