EMAIL THIS PAGE TO A FRIEND
Stem Cells and Development
Oxidative stress increases neurogenesis and oligodendrogenesis in adult neural progenitor cells.
PMID 24773127
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7942
Sirtinol, ≥95% (HPLC)
C26H22N2O2