EMAIL THIS PAGE TO A FRIEND
Anticancer research
Levonorgestrel-impregnated Intrauterine device reduces occurrence of hyperplastic polyps: a population-based follow-up cohort study.
PMID 24778038
Abstract
The main objective was to investigate if occurrence of hyperplastic polyps was reduced by use of the levonorgestrel-impregnated intrauterine system (LNG-IUS, Mirena®; Bayer) and if the LNG-IUS was more effective compared to oral medroxyprogesterone acetate (MPA) or observation-only. Patients (N=59) with hyperplastic polyps were given LNG-IUS, 10 mg oral MPA taken 10 days per cycle, or had observation-only for six months. Diagnosis of histological specimens was performed by light microscopy according to the WHO classification and D-score prior to and after six months therapy. No polyps were found in women treated with LNG-IUS (18/18). Five women treated with cyclic MPA (5/20, 25%) and two (2/21, 9%) with observation had normal endometrium without polyps after six months. No former study has shown that LNG-IUS is effective at reducing the occurrence of hyperplastic endometrial polyps. The effect is superior to that of oral progestin and observation-only.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1362602
Levonorgestrel, United States Pharmacopeia (USP) Reference Standard
C21H28O2
Y0001618
Levonorgestrel for system suitability 2, European Pharmacopoeia (EP) Reference Standard
C21H28O2
20357
ORBO 2-HMP on Supelpak 20N Tube, W,W,W separators, O.D. × L 8 mm × 115 mm, pkg of 25 ea
20257-U
ORBO 23 2-HMP on Amberlite® XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
20231
ORBO 24 2-HMP on Amberlite® XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
20832-U ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20034 ORBO 403 Tenax® TA specially treated (60/80), 100/50 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 50 ea
20262 ORBO 42 Small Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 66/33 mg, W,F,F (specially treated) separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20258 ORBO 43 Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20260-U ORBO 44Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20350
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
20030-U
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20032
ORBO 506 Activated Silica Gel (45/60) 300/150 mg, W,W,W separators, O.D. × L 8 mm × 75 mm, pkg of 50 ea
20263
ORBO 52 Large Activated Silica Gel (45/60) 150/150 mg, W,W,W separators, O.D. × L 8 mm × 75 mm, pkg of 50 ea
20229
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20265
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
20033
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20049 ORBO 605 Amberlite® XAD®-2 specially cleaned (20/50) 100/50 mg, W,W,W separators, O.D. × L 6 mm × 100 mm, pkg of 50 ea
20050-U ORBO 608 Amberlite® XAD®-2 (20/50) 150/75 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
20051 ORBO 609 Amberlite® XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
20052 ORBO 613 Amberlite® XAD®-4 80/40 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20029-U ORBO 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
20812-U ORBO 80 Coated Filter, with cassettes (unassembled)
20811
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
20069
ORBO 827 LpDNPH Coated Filter, 37mm, pkg of 25 ea
20362 ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate