EMAIL THIS PAGE TO A FRIEND
Journal of Cellular and Molecular Medicine
Bone marrow mesenchymal stem cells protected post-infarcted myocardium against arrhythmias via reversing potassium channels remodelling.
PMID 24780005
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0355000
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
C5H9NO4