EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Evaluation of ¹³C- and ²H-labeled internal standards for the determination of amphetamines in biological samples, by reversed-phase ultra-high performance liquid chromatography-tandem mass spectrometry.
PMID 24780257
Abstract
Stable isotope-labeled internal standards (SIL-ISs) are often used when applying liquid chromatography-tandem mass spectrometry (LC-MS/MS) to analyze for legal and illegal drugs. ISs labeled with (13)C, (15)N, and (18)O are expected to behave more closely to their corresponding unlabeled analytes, compared with that of the more classically used (2)H-labeled ISs. This study has investigated the behavior of amphetamine, (2)H3-, (2)H5, (2)H6-, (2)H8-, (2)H11-, and (13)C6-labeled amphetamine, during sample preparation by liquid-liquid extraction and LC-MS/MS analyses. None or only minor differences in liquid-liquid extraction recoveries of amphetamine and the SIL-ISs were observed. The chromatographic resolution between amphetamine and the (2)H-labeled amphetamines increased with the number of (2)H-substitutes. For chromatographic studies we also included seven additional (13)C6-amphetamines and their analytes. All the (13)C6-labeled ISs were co-eluting with their analytes, both when a basic and when an acidic mobile phase were used. MS/MS analyses of amphetamine and its SIL-ISs showed that the ISs with the highest number of (2)H-substitutes required more energy for fragmentation in the collision cell compared with that of the ISs with a lower number. The findings, in this study, support those of previous studies, showing that (13)C-labeled ISs are superior to (2)H-labeled ISs, for analytical purposes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A-007
(±)-Amphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C9H13N
A-011
(±)-Amphetamine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material
C9H13N
09684
Ammonia, puriss., anhydrous, ≥99.9%
H3N
09735
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
CH5NO2
70221
Ammonium formate, for mass spectrometry, ≥99.0%
CH5NO2
227048
Cyclohexane, anhydrous, 99.5%
C6H12
650455
Cyclohexane, for HPLC, ≥99.9%
C6H12
34855
Cyclohexane, for HPLC, ≥99.7%
C6H12
33117
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
C6H12
179191
Cyclohexane, ACS reagent, ≥99%
C6H12
676861
Cyclohexane, ACS reagent, ≥99%
C6H12
05-7040
Cyclohexane, JIS special grade
C6H12
05-7070
Cyclohexane, for HPLC
C6H12
C100307
Cyclohexane, Laboratory Reagent, ≥99.8%
C6H12
16355
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
HNO3
306975
tert-Butyl methyl ether, anhydrous, 99.8%
C5H12O
675407
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
C5H12O
08603
tert-Butyl methyl ether, analytical standard
C5H12O
293210
tert-Butyl methyl ether, HPLC grade, for HPLC, 99.8%
C5H12O
PHR1560
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
C5H12O