EMAIL THIS PAGE TO A FRIEND
Experimental Lung Research
Tiotropium effects on airway inflammatory events in the cat as an animal model for acute cigarette smoke-induced lung inflammation.
PMID 24784973