EMAIL THIS PAGE TO A FRIEND
Obstetrics and gynecology
What is new in obstetric antecedents of chronic disease? Best articles from the past year.
PMID 24785618
Abstract
This month, we focus on current research in obstetric antecedents of chronic disease. Dr Malone discusses four recent publications, and each is concluded with a "bottom line" that is the take-home message. The complete reference for each can be found in on this page along with direct links to the abstracts.