EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Comparison of a novel TiO?/diatomite composite and pure TiO? for the purification of phosvitin phosphopeptides.
PMID 24786220
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277908
Ammonium molybdate, 99.98% trace metals basis
H8MoN2O4
A1343
Ammonium molybdate, meets USP testing specifications
H24Mo7N6O24 · 4H2O
A7302
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
M0878
Ammonium molybdate tetrahydrate, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
M1019
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
H24Mo7N6O24 · 4H2O
09878
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
H24Mo7N6O24 · 4H2O
C8027
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
CuO4S · 5H2O
C3036
Copper(II) sulfate pentahydrate, plant cell culture tested, ≥98%
CuO4S · 5H2O
203165
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
CuO4S · 5H2O
469130
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
CuO4S · 5H2O
209198
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
CuO4S · 5H2O
C7631
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
CuO4S · 5H2O
31293
Copper(II) sulfate pentahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%
CuO4S · 5H2O
61245
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
CuO4S · 5H2O
12849
Copper(II) sulfate pentahydrate, puriss., suitable for, meets analytical specification of Ph. Eur., BP, USP,FCC
CuO4S · 5H2O
RES10395-B7
Cupric sulfate pentahydrate, PharmaGrade, EP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
CuO4S · 5H2O
H9003
Hydroquinone, ReagentPlus®, ≥99%
C6H6O2
H3660
Hydroquinone, meets USP testing specifications
C6H6O2
74347
Hydroquinone, certified reference material, TraceCERT®
C6H6O2
1324002
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
C6H6O2
H17902
Hydroquinone, ReagentPlus®, ≥99.5%
C6H6O2
PHR1469
Hydroquinone, Pharmaceutical Secondary Standard; Certified Reference Material
C6H6O2
S0505
Sodium sulfite, ≥98%
Na2O3S
71988
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)
Na2O3S
31454
Sodium sulfite, puriss. p.a., Reag. Ph. Eur., anhydrous, 98-100.0% (iodometric)
Na2O3S
13471
Sodium sulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, anhydrous, 95.0-100.0% (iodometric)
Na2O3S
08981
Sodium sulfite, analytical standard
Na2O3S
239321
Sodium sulfite, ACS reagent, ≥98.0%
Na2O3S
207845
Sodium sulfite, reagent grade, 98%
Na2O3S
S4672
Sodium sulfite, BioXtra, ≥98%
Na2O3S