EMAIL THIS PAGE TO A FRIEND
The international journal of neuropsychopharmacology
Chronic fluoxetine treatment alters the structure, connectivity and plasticity of cortical interneurons.
PMID 24786752
Abstract
Novel hypotheses suggest that antidepressants, such as the selective serotonin reuptake inhibitor fluoxetine, induce neuronal structural plasticity, resembling that of the juvenile brain, although the underlying mechanisms of this reopening of the critical periods still remain unclear. However, recent studies suggest that inhibitory networks play an important role in this structural plasticity induced by fluoxetine. For this reason we have analysed the effects of a chronic fluoxetine treatment in the hippocampus and medial prefrontal cortex (mPFC) of transgenic mice displaying eGFP labelled interneurons. We have found an increase in the expression of molecules related to critical period plasticity, such as the polysialylated form of the neural cell adhesion molecule (PSA-NCAM), GAD67/65 and synaptophysin, as well as a reduction in the number of parvalbumin expressing interneurons surrounded by perineuronal nets. We have also described a trend towards decrease in the perisomatic inhibitory puncta on pyramidal neurons in the mPFC and an increase in the density of inhibitory puncta on eGFP interneurons. Finally, we have found that chronic fluoxetine treatment affects the structure of interneurons in the mPFC, increasing their dendritic spine density. The present study provides evidence indicating that fluoxetine promotes structural changes in the inhibitory neurons of the adult cerebral cortex, probably through alterations in plasticity-related molecules of neurons or the extracellular matrix surrounding them, which are present in interneurons and are known to be crucial for the development of the critical periods of plasticity in the juvenile brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405553
Anti-CAMK2A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2500435
Anti-FYN antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA023887
Anti-FYN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100865
Anti-FYN antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2500752
Anti-Parvalbumin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA048536
Anti-PVALB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502906
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
S9075
Anti-Synemin (N-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
S8950
Anti-Synemin antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA002858
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102352
Anti-SYP antibody produced in rabbit, affinity isolated antibody
WH0000815M1
Monoclonal Anti-CAMK2A antibody produced in mouse, clone 2C4, purified immunoglobulin, buffered aqueous solution
SAB1403629
Monoclonal Anti-CAMK2A antibody produced in mouse, clone 2C12, purified immunoglobulin, buffered aqueous solution
WH0023336M3
Monoclonal Anti-DMN antibody produced in mouse, clone 4G5, purified immunoglobulin, buffered aqueous solution
WH0002534M3
Monoclonal Anti-FYN antibody produced in mouse, clone 1A3, purified immunoglobulin, buffered aqueous solution
SAB1403844
Monoclonal Anti-FYN antibody produced in mouse, clone 3G11-F9, purified immunoglobulin, buffered aqueous solution
SAB4200545
Monoclonal Anti-Parvalbumin antibody produced in mouse, ~1.0 mg/mL, clone PARV-19, purified immunoglobulin
A2795
N-Acetyl-D-galactosamine, ~98%
C8H15NO6