EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
What is the right dose of systemic corticosteroids for intensive care unit patients with chronic obstructive pulmonary disease exacerbations? A question in search of a definitive answer.
PMID 24787061