EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
NanoClusters surface area allows nanoparticle dissolution with microparticle properties.
PMID 24788354
Abstract
Poorly water-soluble drugs comprise the majority of new drug molecules. Nanoparticle agglomerates, called NanoClusters, can increase the dissolution rate of poorly soluble compounds by increasing particle surface area. Budesonide and danazol, two poorly soluble steroids, were studied as model compounds. NanoCluster suspensions were made using a Netzsch MiniCer media mill with samples collected between 5 and 15 h and lyophilized. Differential scanning calorimetry (DSC) and powder X-ray Diffraction were used to evaluate the physicochemical properties of the powders, and Brunauer, Emmett and Teller (BET) analysis was used to determine surface area. Scanning electron microscopy confirmed NanoClusters were between 1 and 5 μm. NanoCluster samples showed an increase in dissolution rate compared with the micronized stock and similar to a dried nanoparticle suspension. BET analysis determined an increase in surface area of eight times for budesonide NanoClusters and 10-15 times for danazol NanoClusters compared with the micronized stock. Melting temperatures decreased with increased mill time of NanoClusters by DSC. The increased surface area of NanoClusters provides a potential micron-sized alternative to nanoparticles to increase dissolution rate of poorly water-soluble drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262994
Yttrium, chips, 99.9% trace rare earth metals basis
Y
451347
Yttrium, dendritic pieces, 99.9% trace metals basis
Y
261319
Yttrium, ingot, 99.9% trace rare earth metals basis
Y
261327
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Y
GF09119279
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Y
GF77674719
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Y
GF11301398
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Y
GF86394156
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Y
GF91377478
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Y
GF87149481
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Y
GF03037148
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Y
GF55118257
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Y
GF33100298
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Y
GF75315719
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Y
GF61507740
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Y
GF40879008
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Y
GF12674617
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Y
GF84327037
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Y
GF68048860
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Y
GF68472692
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Y
GF35512725
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Y
GF95617020
Yttrium, wire reel, 0.05m, diameter 1.0mm, square section, 99.9%
Y
GF02763963
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Y
GF39091158
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Y
GF79949255
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Y
773972
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Y