EMAIL THIS PAGE TO A FRIEND
Archives of virology
Epidemiology of rotavirus diarrhoea in Albania.
PMID 24788847
Abstract
The aim of the study was to estimate the prevalence of rotavirus disease in children<5 years old in Tirana, Albania, and to monitor and characterize the rotavirus genotypes. Rotavirus was detected in 21% of samples, more frequently in children under 2 years of age, which accounted for 80.8% of all positive cases. Among all rotavirus-positive samples collected, G4P[8] was the most prevalent genotype (38%), followed by G1P[8] (36.6%). The use of safe and effective rotavirus vaccines for the prevention of severe diarrhoea and the reduction of treatment costs will be of great importance for Albania.