EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
A capillary electrophoresis procedure for the screening of oligosaccharidoses and related diseases.
PMID 24788891
Abstract
The most widely used method for the biochemical screening of oligosaccharidoses is the analysis of the urinary oligosaccharide pattern by thin-layer chromatography on silica gel plates. However, this method is not always sensitive enough, and it is extremely time-consuming and laborious. In this work, the analysis of the urine oligosaccharide pattern was standardized for the first time by using capillary electrophoresis with laser-induced fluorescence (CE-LIF) detection (Beckman P/ACE MDQ) with a 488-nm argon ion laser module. All of the analyses were conducted using the Carbohydrate Labeling and Analysis Kit (Beckman-Coulter), which derivatizes samples with 8-aminopyrene-1,3,6-trisulfonate. Urine samples from 40 control subjects (age range, 1 week to 16 years) and from ten patients diagnosed with eight different lysosomal diseases (six of them included in the Educational Oligosaccharide Kit from ERNDIM EQA schemes) were analyzed. Two oligosaccharide excretion patterns were established in our control population according to age (younger or older than 1 year of age). Abnormal peaks with slower migration times than the tetrasaccharide position were observed for fucosidosis, α-mannosidosis, GM1 gangliosidosis, GM2 gangliosidosis variant 0, Pompe disease, and glycogen storage disease type 3. In conclusion, the first CE-LIF method to screen for oligosaccharidoses and related diseases, which also present oligosacchariduria, has been standardized. In all of the cases, the urine oligosaccharide analysis was strongly informative and showed abnormal patterns that were not present in any of the urine samples from the control subjects. Only urine from patients with aspartylglucosaminuria and Schindler disease displayed normal results.