PloS one
Geminin overexpression promotes imatinib sensitive breast cancer: a novel treatment approach for aggressive breast cancers, including a subset of triple negative.
PMID 24789045
Abstract
Breast cancer is the second leading cause of cancer-related deaths in women. Triple negative breast cancer (TNBC) is an aggressive subtype that affects 10-25% mostly African American women. TNBC has the poorest prognosis of all subtypes with rapid progression leading to mortality in younger patients. So far, there is no targeted treatment for TNBC. To that end, here we show that c-Abl is one of several tyrosine kinases that phosphorylate and activate geminin's ability to promote TNBC. Analysis of >800 breast tumor samples showed that geminin is overexpressed in ∼50% of all tumors. Although c-Abl is overexpressed in ∼90% of all tumors, it is only nuclear in geminin overexpressing tumors. In geminin-negative tumors, c-Abl is only cytoplasmic. Inhibiting c-Abl expression or activity (using imatinib or nilotinib) prevented geminin Y150 phosphorylation, inactivated the protein, and most importantly converted overexpressed geminin from an oncogene to an apoptosis inducer. In pre-clinical orthotopic breast tumor models, geminin-overexpressing cells developed aneuploid and invasive tumors, which were suppressed when c-Abl expression was blocked. Moreover, established geminin overexpressing orthotopic tumors regressed when treated with imatinib or nilotinib. Our studies support imatinib/nilotonib as a novel treatment option for patients with aggressive breast cancer (including a subset of TNBCs)-overexpressing geminin and nuclear c-Abl.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-570
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker is a Rabbit Polyclonal Antibody for detection of Histone H3 phosphorylated at serine 10.This highly published Ab, also known as Anti-H3S10p, has been validated in ICC, IP & WB.
AV46350
Anti-CDT1 antibody produced in rabbit, affinity isolated antibody
SAB2701049
Anti-CDT1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306180 ANTI-PHOSPHO-H3(S10) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV37192
Anti-SP1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
S9809 Anti-Sp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001853
Anti-SP1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV43585
Anti-SP1 antibody produced in rabbit, affinity isolated antibody
HPA012292
Anti-SP1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502836
Anti-SP1 antibody produced in rabbit, affinity isolated antibody
EHU141291 MISSION® esiRNA, esiRNA human ABL1 (esiRNA1)
EMU071391 MISSION® esiRNA, esiRNA targeting mouse Abl1 (esiRNA1)
A5844 Monoclonal Anti-c-Abl antibody produced in mouse, clone ABL-148, ascites fluid
WH0051053M1
Monoclonal Anti-GMNN antibody produced in mouse, clone 1A8, purified immunoglobulin, buffered aqueous solution
PLA0044
Rabbit anti-Sp1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.