EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Diabetes, aging, and their tissue complications.
PMID 24789881
Abstract
The inactivation of NO by advanced glycation endproducts (AGEs), which accumulate on tissue proteins as a function of age and hyperglycemia, focused attention on the role of these ubiquitous posttranslational modifications in acquired impairments of vascular reactivity and other signaling processes. This observation occurred during a watershed period of basic and translational research in glycation that encompassed new pathologic phenomena and novel intervention strategies. How has the AGE paradigm for the tissue complications of aging and diabetes fared since the identification of the link between these glycation products and NO inactivation, and what lessons may be offered for future investigations?