EMAIL THIS PAGE TO A FRIEND
Journal of Analytical Toxicology
Quantitative analysis of zopiclone, N-desmethylzopiclone, zopiclone N-oxide and 2-amino-5-chloropyridine in urine using LC-MS-MS.
PMID 24790062