EMAIL THIS PAGE TO A FRIEND
Arteriosclerosis, thrombosis, and vascular biology
Targeted delivery of pulmonary arterial endothelial cells overexpressing interleukin-8 receptors attenuates monocrotaline-induced pulmonary vascular remodeling.
PMID 24790141
Abstract
Interleukin-8 (IL-8) receptors IL8RA and IL8RB (IL8RA/B) on neutrophil membranes bind to IL-8 with high affinity and play a critical role in neutrophil recruitment to sites of injury and inflammation. This study tested the hypothesis that administration of rat pulmonary arterial endothelial cells (ECs) overexpressing IL8RA/B can accelerate the adhesion of ECs to the injured lung and inhibit monocrotaline-induced pulmonary inflammation, arterial thickening and hypertension, and right ventricular hypertrophy. The treatment groups included 10-week-old ovariectomized Sprague-Dawley rats that received subcutaneous injection of PBS (vehicle), a single injection of monocrotaline (monocrotaline alone, 60 mg/kg, SC), monocrotaline followed by intravenous transfusion of ECs transduced with the empty adenoviral vector (null-EC), and monocrotaline followed by intravenous transfusion of ECs overexpressing IL8RA/B (1.5 × 10(6) cells/rat). Two days or 4 weeks after monocrotaline treatment, endothelial nitric oxide synthase, inducible nitric oxide synthase, cytokine-induced neutrophil chemoattractant-2β (IL-8 equivalent in rat), and monocyte chemoattractant protein-1 expression, neutrophil and macrophage infiltration into pulmonary arterioles, and arteriolar and alveolar morphology were measured by histological and immunohistochemical techniques. Proinflammatory cytokine/chemokine protein levels were measured by Multiplex rat-specific magnetic bead-based sandwich immunoassay in total lung homogenates. Transfusion of ECs overexpressing IL8RA/B significantly reduced monocrotaline-induced neutrophil infiltration and proinflammatory mediator (IL-8, monocyte chemoattractant protein-1, inducible nitric oxide synthase, cytokine-induced neutrophil chemoattractant, and macrophage inflammatory protein-2) expression in lungs and pulmonary arterioles and alveoli, pulmonary arterial pressure, and pulmonary arterial and right ventricular hypertrophy and remodeling. These provocative findings suggest that targeted delivery of ECs overexpressing IL8RA/B is effective in repairing the injured pulmonary vasculature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2401
Crotaline
C16H23NO6