EMAIL THIS PAGE TO A FRIEND
Environment international
Establishing the importance of human health risk assessment for metals and metalloids in urban environments.
PMID 24791693
Abstract
Rapid development, industrialisation, and urbanisation have resulted in serious contamination of soil by metals and metalloids from anthropogenic sources in many areas of the world, either directly or indirectly. Exponential urban and economic development has resulted in human populations settling in urban areas and as a result being exposed to these pollutants. Depending on the nature of the contaminant, contaminated urban soils can have a deleterious effect on the health of exposed populations and may require decontamination, recovery, remediation and restoration. Therefore, human health risk assessments in urban environments are very important. In the case of Spain, there are few studies regarding risk assessment of trace elements in urban soils, and those that exist have been derived mainly from areas potentially exposed to industrial contamination or in the vicinity of point pollution. The present study analysed Al, As, Be, Cd, Cr, Cu, Hg, Mn, Ni, Pb, Sn, Ti, Tl, V and Zn soil concentrations in and around the city of Alcalá de Henares (35 km NE of Madrid). Soil samples were collected in public parks and recreation areas within the city and in an industrial area on the periphery of the city. From these results, an assessment of the health risk for the population was performed following the methodology described by the US EPA (1989). In general, it was observed that there could be a potential increased risk of developing cancer over a lifetime from exposure to arsenic (As) through ingestion of the soils studied (oral intake), as well as an increased risk of cancer due to inhalation of chromium (Cr) present in re-suspended soils from the industrial area. Our group has previously reported (Granero and Domingo, 2002; Peña-Fernández et al., 2003) that there was an increased risk of developing cancer following exposure to As in the same soils in a previous study. Therefore, it is necessary to reduce the levels of contaminants in these soils, especially As and Cr as these have been found to exceed safe levels for human health.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF07622759
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Cr
GF35390651
Chromium, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF46122645
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF31792956
Chromium, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF91240994
Chromium, foil, not light tested, 25x25mm, thickness 0.075mm, permanent polyester support, 99.99+%
Cr
GF98190861
Chromium, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF76896400
Chromium, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF64373766
Chromium, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF34299748
Chromium, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF31161754
Chromium, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF91709196
Chromium, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr