EMAIL THIS PAGE TO A FRIEND
Steroids
Heterocyclic ring extension of androstenedione: synthesis and cytotoxicity of fused pyran, pyrimidine and thiazole derivatives.
PMID 24793334
Abstract
The reaction of androstenedione with either malononitrile or ethyl cyanoacetate and aromatic aldehydes 2a-c gave the pyran derivatives 4a-f, respectively. On the other hand, the reaction of androstenedione with thiourea and the aromatic aldehydes 2a-c gave the pyrimidine derivatives 6a-c, respectively. Compound 6b reacted with 2-bromo-1-arylethanone derivatives 7a-d to give the indeno[2,1-e]thiazole derivatives 8a-d. Some of the produced compounds were used for further heterocyclization reactions. The cytotoxicity of the newly obtained products was evaluated against some cancer cell lines and a normal cell line.